Learn More
PYREX™ Disposable Rimless Flat-Bottom Glass Tubes
Ideal for plant tissue culture and general culture work
Specifications
| Material | Borosilicate Glass |
|---|---|
| Shape | Flat |
| Disposable | No |
| For Use With | For tissue culture work and general bacteriological use |
| Label Type | None |
Description
These PYREX™ flat bottom reusable culture tubes are ideal for plant tissue culture and general culture work. The flat bottom facilitates use with agar slants. Walls and bottoms are of uniform thickness. The tubes are rimless for greater convenience in plugging and rack storage. Optional polypropylene two position closures (Nos. 26794-25 or 26795-25) can be opened for gas exchange or closed for humidified environment. Internal drip ring minimizes contamination.Order Info
Supplied without closures; except Pyrex 14-957-85E, -F, -G, and -K caps (sold separately).
Specifications
| Borosilicate Glass | |
| No | |
| None | |
| Non-sterile |
| Flat | |
| For tissue culture work and general bacteriological use | |
| No | |
| No |