Learn More
PYREX™ Culture Flask Closure
For use with 1135 culture flasks, 1138/10 Fernbach flasks and 1141 baffled flasks.
£52.00 - £172.00
Product Code | Brand | Closure Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Material | Price | Quantity & Availability | |||||
12524917
|
Pyrex
4540/38P |
Polypropylene |
£52.00
Pack of 10 |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
12687266
|
Pyrex
4550/38M |
Aluminum |
£172.00
Pack of 10 |
Estimated Shipment: 26-04-2024
Log in to see stock. |
|||||
Description
- Two types of closure - Anodised aluminum - Two-position polypropylene
- Suitable for autoclaving
Specifications
10 | |
Autoclavable |