Learn More
Pyrex™ Borosilicate Glass Vacuum Desiccator Cover
Product Code. 11985869
Change view
Click to view available options
For Use With (Equipment):
Vacuum desiccators
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
11985869 Vacuum desiccators Each
1 options
Product Code. 11985869 Supplier Pyrex Supplier No. 1594/04D
  • ONLINE EXCLUSIVE PRICE
    £393.67
    £256.45 / Pack of 1
    Save £137.22
    34% Off
Estimated Shipment: 06-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed for use under vacuum.
Designed for use under vacuum.
  • Designed for use under vacuum
  • Sturdy construction in borosilicate glass
  • Ground glass flanges
  • Cover has a 24/29 socket to accept desiccator stopcocks
  • Accommodates 200mm diameter plates
  • Plates are positioned on an internal ledge within the base
  • Accessory metal perforated plate and Rotaflo™ glass/PTFE stopcock available
Specifications
Diameter (Metric) Outer 220 mm
Product Type Vacuum Desiccator Cover
Material Borosilicate Glass
For Use With (Equipment) Vacuum desiccators