missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Borosilicate Glass Reagent Bottles with Polypropylene Cap and Pouring Ring

Product Code. 15727927
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
15,000 mL
150 mL
20,000 mL
2000 mL
25 mL
250 mL
3500 mL
50 mL
500 mL
5000 mL
Unit Size:
Each
Pack of 10
This item is not returnable. View return policy

Product Code. 15727927

Brand: Pyrex 1516/05D

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Mix and store a wide range of culture mediums with these glass reagent bottles. Pyrex™ Borosilicate Glass Reagent Bottles with Polypropylene Cap and Pouring Ring feature a high chemical resistance and are designed with sloped shoulders for smooth pouring.

Mix and store a wide range of culture mediums with these glass reagent bottles. PyrexTM Borosilicate Glass Reagent Bottles with Polypropylene Cap and Pouring Ring feature a high chemical resistance and are designed with sloped shoulders for smooth pouring.

  • Bottles complete with polypropylene cap and drip-free pouring ring
  • Complies with ISO 4796
  • Printed with Trace Code for downloadable batch certificate
  • Sloping shoulders for smooth pouring action
  • White graduations and marking spot
  • Manufactured from temperature and chemically resistant borosilicate glass
  • For use up to 140°C
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 150 mL
Capacity (English) 5.1 oz.
Color Clear
Diameter (English) 2.4 in.
For Use With (Application) Ideal for mixing and storage of culture medium
Material Borosilicate Glass
Quantity 10 Pk.
Thread Size (Metric) GL 45
Certifications/Compliance ISO 4796
Closure Included Yes
Disposable Reusable
Graduated Yes
Mouth Wide
Shape Round
Height (Metric) 105 mm
Product Type Reagent Bottle
For Use With (Equipment) Ideal for Mixing and Storage of Culture Medium
Height (English) 4.1 in.
Closure Material Polypropylene
Closure Size 45 mm
Diameter (Metric) 62 mm
Closure Type Screw Cap
Autoclavable Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.