missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Borosilicate Glass Reagent Bottles with Polypropylene Cap and Pouring Ring
Description
Mix and store a wide range of culture mediums with these glass reagent bottles. PyrexTM Borosilicate Glass Reagent Bottles with Polypropylene Cap and Pouring Ring feature a high chemical resistance and are designed with sloped shoulders for smooth pouring.
- Bottles complete with polypropylene cap and drip-free pouring ring
- Complies with ISO 4796
- Printed with Trace Code for downloadable batch certificate
- Sloping shoulders for smooth pouring action
- White graduations and marking spot
- Manufactured from temperature and chemically resistant borosilicate glass
- For use up to 140°C
Specifications
Specifications
| Capacity (Metric) | 10,000 mL |
| Capacity (English) | 2.6 gal. |
| Color | Clear |
| Diameter (English) | 8.9 in. |
| For Use With (Application) | Ideal for mixing and storage of culture medium |
| Includes | Polypropylene screw cap and pouring ring |
| Material | Borosilicate Glass |
| Quantity | 1 Ea. |
| Thread Size (Metric) | GL 45 |
| Certifications/Compliance | ISO 4796 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?