missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Borosilicate Glass Reagent Bottles with Polypropylene Cap and Pouring Ring

Product Code. 11972619
Change view
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
15,000 mL
150 mL
20,000 mL
2000 mL
25 mL
250 mL
3500 mL
50 mL
500 mL
5000 mL
Unit Size:
Each
Pack of 10
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11972619 50 mL Pack of 10
11962619 25 mL Pack of 10
11982619 100 mL Pack of 10
15727927 150 mL Pack of 10
11992619 250 mL Pack of 10
11902629 500 mL Pack of 10
11912629 1000 mL Pack of 10
11922629 2000 mL Pack of 10
15747927 3500 mL Each
12094637 5000 mL Each
12094717 10,000 mL Each
10670191 15,000 mL Each
12643796 20,000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 11972619 Supplier Pyrex Supplier No. 1516/02D
Discount Lab Supplies
Use promo code 28170
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

Mix and store a wide range of culture mediums with these glass reagent bottles. Pyrex™ Borosilicate Glass Reagent Bottles with Polypropylene Cap and Pouring Ring feature a high chemical resistance and are designed with sloped shoulders for smooth pouring.

Mix and store a wide range of culture mediums with these glass reagent bottles. PyrexTM Borosilicate Glass Reagent Bottles with Polypropylene Cap and Pouring Ring feature a high chemical resistance and are designed with sloped shoulders for smooth pouring.

  • Bottles complete with polypropylene cap and drip-free pouring ring
  • Complies with ISO 4796
  • Printed with Trace Code for downloadable batch certificate
  • Sloping shoulders for smooth pouring action
  • White graduations and marking spot
  • Manufactured from temperature and chemically resistant borosilicate glass
  • For use up to 140°C

Specifications

Capacity (Metric) 50 mL
Color Clear
Diameter (English) 1.8 in.
For Use With (Application) Ideal for mixing and storage of culture medium
Includes Polypropylene screw cap and pouring ring
Material Borosilicate Glass
No. per Pack 10/PK
Quantity 10 Pk.
Thread Size (Metric) GL 32
Closure Included Yes
Graduated Yes
Mouth Wide
Height (Metric) 88 mm
For Use With (Equipment) Ideal for Mixing and Storage of Culture Medium
Height (English) 3.5 in.
Closure Material Polypropylene
Closure Size 32 mm
Diameter (Metric) 46 mm
Closure Type Screw Cap
Autoclavable Autoclavable
Capacity (English) 1.7 oz.
Shape Round
Certifications/Compliance ISO 4796
Disposable Reusable
Product Type Reagent Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.