missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Borosilicate Glass Reagent Bottles

Product Code. 11357854
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
15,000 mL
20,000 mL
2000 mL
25 mL
250 mL
50 mL
500 mL
5000 mL
Unit Size:
Each
Pack of 10
Pack of 5
This item is not returnable. View return policy

Product Code. 11357854

Brand: Pyrex 1515/18D

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Store reagents, analytical standards and culture media in these borosilicate glass reagent bottles. Pyrex™ Borosilicate Glass Reagent Bottles withstand hot air sterilization up to 180°C, and have excellent chemical resistance.

Store reagents, analytical standards and culture media in these borosilicate glass reagent bottles. PyrexTM Borosilicate Glass Reagent Bottles withstand hot air sterilization up to 180°C, and have excellent chemical resistance.

  • Complies with ISO 4796-1
  • Manufactured from Pyrex™ borosilicate glass
  • Excellent resistance to chemical attack and high temperatures
  • Supplied with PBT screw caps that have a PTFE faced sealing disc
  • Autoclavable
  • White graduations and marking spot
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 10,000 mL
For Use With (Application) Ideal for storage of reagents, analytical standards and culture media etc
Material Borosilicate Glass
No. per Pack 1/PK
Closure Color Black
Certifications/Compliance ISO 4796-1
Closure Included Yes
Graduated Yes
Height (Metric) 395 mm
Closure Liner PTFE
Closure Material Phenolic
Closure Size 60 mm
Diameter (Metric) 227 mm
Closure Type Screw Cap
Autoclavable Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.