Learn More
Pyrex™ Borosilicate Glass Fernbach Flask with Screw Cap
Brand: Pyrex 1139/10
This item is not returnable.
View return policy
Description
Specifications
2800 mL | |
Preparation of cultures | |
210 mm |
Borosilicate Glass | |
GL 45 |