missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrethroid hydrolase Ces2e Antibody, mFluor Violet 500 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Pyrethroid hydrolase Ces2e Polyclonal antibody specifically detects Pyrethroid hydrolase Ces2e in Mouse samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | Pyrethroid hydrolase Ces2e |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 500 SE |
| Dilution | Western Blot, ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 9030624L02Rik, ACH M1;CE-2;CES 2;CES2;CES2A1;ces2A3;Ces2c;Ces2e;ICE;PCE-2, carboxylesterase 5, carboxylic ester hydrolase, Ces5 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Mouse Pyrethroid hydrolase Ces2e (Accession#: NP_766347.1; Met1-His556) |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?