missing translation for 'onlineSavingsMsg'
Learn More

Pyrethroid hydrolase Ces2e Antibody, mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18416629 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416629 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416629 Supplier Novus Biologicals Supplier No. NBP306082MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Pyrethroid hydrolase Ces2e Polyclonal antibody specifically detects Pyrethroid hydrolase Ces2e in Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pyrethroid hydrolase Ces2e
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias 9030624L02Rik, ACH M1;CE-2;CES 2;CES2;CES2A1;ces2A3;Ces2c;Ces2e;ICE;PCE-2, carboxylesterase 5, carboxylic ester hydrolase, Ces5
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Mouse Pyrethroid hydrolase Ces2e (Accession#: NP_766347.1; Met1-His556)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 234673
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.