missing translation for 'onlineSavingsMsg'
Learn More

Pyrethroid hydrolase Ces2e Antibody, CoraFluor™ 1, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP3-06082CL1

Product Code. 30153814

  • £622.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Pyrethroid hydrolase Ces2e Polyclonal antibody specifically detects Pyrethroid hydrolase Ces2e in Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pyrethroid hydrolase Ces2e
Polyclonal
PBS
Rabbit
Antigen and protein A Affinity-purified
RUO
Primary
Mouse
Purified
ELISA, Western Blot
CoraFluor 1
9030624L02Rik, ACH M1;CE-2;CES 2;CES2;CES2A1;ces2A3;Ces2c;Ces2e;ICE;PCE-2, carboxylesterase 5, carboxylic ester hydrolase, Ces5
Produced in rabbits immunized with purified, recombinant Mouse Pyrethroid hydrolase Ces2e (Accession#: NP_766347.1; Met1-His556)
0.1 mL
Immunology
234673
Store at 4°C in the dark. Do not freeze.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.