missing translation for 'onlineSavingsMsg'
Learn More

Pyrazolo[1,5-a]pyrimidine-5-carboxylic Acid, TRC

Product Code. 30422149
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
Product Code. Quantity unitSize
30422149 250 mg 250mg
30416720 25 mg -
30413926 50 mg -
3 options
This item is not returnable. View return policy

Product Code. 30422149

Brand: Toronto Research Chemicals TRCP846535250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1086375-50-9
Formula Weight 163.0382
InChI Formula InChI=1S/C7H5N3O2/c11-7(12)5-2-4-10-6(9-5)1-3-8-10/h1-4H,(H,11,12)
IUPAC Name pyrazolo[1,5-a]pyrimidine-5-carboxylic acid
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.13
SMILES OC(=O)c1ccn2nccc2n1
Chemical Name or Material Pyrazolo[1,5-a]pyrimidine-5-carboxylic Acid

Specifications

Quantity 250 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.