missing translation for 'onlineSavingsMsg'
Learn More

Pyrazolo[1,5-a]pyrimidine-5-carboxylic acid, 97%

Product Code. p-4228892 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10220270

Brand: Thermo Scientific Acros 432720010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1086375-50-9 | C7H5N3O2 | 163.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1086375-50-9
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.14
MDL Number MFCD09414725
InChI Key SBDWLDBGGVPVAR-UHFFFAOYSA-N
Synonym pyrazolo 1,5-a pyrimidine-5-carboxylic acid, pyrazolo 1?5-a pyrimidine-5-carboxylic acid, 8-hydropyrazolo 1,5-a pyrimidine-5-carboxylic acid
PubChem CID 28875802
IUPAC Name pyrazolo[1,5-a]pyrimidine-5-carboxylic acid
SMILES OC(=O)C1=NC2=CC=NN2C=C1

Specifications

Melting Point 222°C to 225°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 163.14
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Pyrazolo[1, 5-a]pyrimidine-5-carboxylic acid
compliance-icons
Product Identifier
  • Pyrazolo¢1,5-a!pyrimidine-5-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.