missing translation for 'onlineSavingsMsg'
Learn More

Pyrazolo[1,5-a]pyrimidine-5-carboxylic acid, 97%

Product Code. 10078289 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10078289 5 g Glass bottle 5g
10220270 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10078289 Supplier Thermo Scientific Acros Supplier No. 432720050
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1086375-50-9 | C7H5N3O2 | 163.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1086375-50-9
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.14
MDL Number MFCD09414725
InChI Key SBDWLDBGGVPVAR-UHFFFAOYSA-N
Synonym pyrazolo 1,5-a pyrimidine-5-carboxylic acid, pyrazolo 1?5-a pyrimidine-5-carboxylic acid, 8-hydropyrazolo 1,5-a pyrimidine-5-carboxylic acid
PubChem CID 28875802
IUPAC Name pyrazolo[1,5-a]pyrimidine-5-carboxylic acid
SMILES OC(=O)C1=NC2=CC=NN2C=C1

Specifications

Melting Point 222°C to 225°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 163.14
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Pyrazolo[1, 5-a]pyrimidine-5-carboxylic acid
compliance-icons
Product Identifier
  • Pyrazolo[1,5-a]pyrimidine-5-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.