missing translation for 'onlineSavingsMsg'
Learn More

Pyrazolo[1,5-a]pyrimidine-2-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10618550 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10618550 1 g Glass bottle 1g
10536630 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10618550 Supplier Thermo Scientific Acros Supplier No. 434340010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 378211-85-9
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.14
MDL Number MFCD00297221
InChI Key DZPAOAZDQHZRGG-UHFFFAOYSA-N
Synonym pyrazolo 1,5-a pyrimidine-2-carboxylic acid, 8-hydropyrazolo 1,5-a pyrimidine-2-carboxylic acid, 2-pyrazolo 1,5-a pyrimidinecarboxylic acid, pyrazolo 1,5-a pyrimidine-2-carboxylicacid, acmc-1afu0, pyrazolo 1,5-a pyrimidinecarboxylic acid, pyrazolo 1?5-a pyrimidine-2-carboxylic acid, pyrazolo 1,5-a pyrimidine-2-carboxylic acid 9ci, 2-methyl-3-2-trimethylsilyl ethoxy methyl-3h-imidazo 4,5-b pyridin-6-yl boronic acid
PubChem CID 670824
IUPAC Name pyrazolo[1,5-a]pyrimidine-2-carboxylic acid
SMILES C1=CN2C(=CC(=N2)C(=O)O)N=C1

Specifications

Melting Point 289°C to 291°C
Color Orange to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 163.14
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material Pyrazolo[1, 5-a]pyrimidine-2-carboxylic acid
compliance-icons
Product Identifier
  • Pyrazolo¢1,5-a!pyrimidine-2-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.