missing translation for 'onlineSavingsMsg'
Learn More

Pyrazolo[1,5-a]pyridine-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10495122
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10495122

Brand: Thermo Scientific Maybridge CC26501DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 16205-46-2
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00229214
InChI Key HRSDPDBQVZHCRC-UHFFFAOYSA-N
Synonym pyrazolo 1,5-a pyridine-3-carboxylic acid, zlchem 84, 3-pyrazolo 1,5-a pyridinecarboxylic acid, pyrazolo 1,5-a pyridine-3-carboxylicacid, pyrazolo pyridine-3-carboxylic acid, h-pyrazolo 1,5-a pyridine-3-carboxylic acid, 8-hydropyrazolo 1,5-a pyridine-3-carboxylic acid
PubChem CID 2795462
IUPAC Name pyrazolo[1,5-a]pyridine-3-carboxylic acid
SMILES C1=CC2=C(C=NN2C=C1)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 162.15
Percent Purity 97%
Chemical Name or Material Pyrazolo[1, 5-a]pyridine-3-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.