missing translation for 'onlineSavingsMsg'
Learn More

Pyrazole N-Demethyl Sildenafil-d3, TRC

Product Code. 30415586
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30415586 10 mg 10mg
30423943 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30415586 Supplier Toronto Research Chemicals Supplier No. TRCP84280210MG
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185044-03-4
Molecular Formula C21H25D3N6O4S
Molecular Weight (g/mol) 463.57
Synonym 1-[[3-(4,7-Dihydroxy-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl-4- ethoxyphenyl]sulfonyl]-4-(methyl-d3)piperazine, Desmethylsildenafil-d3
SMILES O=S(N1CCN(C([2H])([2H])[2H])CC1)(C2=CC=C(OCC)C(C3=NC(C(CCC)=NN4)=C4C(N3)=O)=C2)=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C21H28N6O4S/c1-4-6-16-18-19(25-24-16)21(28)23-20(22-18)15-13-14(7-8-17(15)31-5-2)32(29,30)27-11-9-26(3)10-12-27/h7-8,13H,4-6,9-12H2,1-3H3,(H,24,25)(H,22,23,28)/i3D3
Formula Weight 463.2081
Chemical Name or Material Pyrazole N-Demethyl Sildenafil-d3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.