missing translation for 'onlineSavingsMsg'
Learn More

Pyrazole-3-boronic acid hydrochloride, 97%, Thermo Scientific™

Product Code. 11996241 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11996241 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11996241 Supplier Thermo Scientific Acros Supplier No. 445510010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1314216-33-5
Assay Percent Range 97%
Packaging Glass Bottle
Molecular Formula C3H5BN2O2·ClH
Quantity 1 g
Synonym 1h-pyrazol-3-yl boronic acid hydrochloride, 2h-pyrazol-3-yl boronic acid hcl, pyrazole-3-boronic acid hydrochloride, 2h-pyrazol-3-yl boronic acid, hcl, 1h-pyrazol-3-ylboronic acid hydrochloride, 2h-pyrazol-3-ylboronic acid hydrochloride, pyrazole-3-boronic acid, hydrochloride, c3h5bn2o2.clh, 2h-pyrazol-3-boronic acid hydrochloride, 1h-pyrazol-5-ylboronic acid-hydrogen chloride 1/1
InChI Key YUVMFPRVYZDAPN-UHFFFAOYSA-N
SMILES B(C1=CC=NN1)(O)O.Cl
IUPAC Name 1H-pyrazol-5-ylboronic acid;hydrochloride
Molecular Weight (g/mol) 148.36
PubChem CID 56776699
Formula Weight 148.36
Percent Purity 97%
Chemical Name or Material Pyrazole-3-boronic acid hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Pyrazole-3-boronic acid hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.