missing translation for 'onlineSavingsMsg'
Learn More

Pyrazinecarboxylic Acid, TRC

Product Code. 30411532
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30411532 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411532 Supplier Toronto Research Chemicals Supplier No. TRCP8407505G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pyrazine-2-carboxylic Acid
CAS 98-97-5
Molecular Formula C5 H4 N2 O2
Quantity 5 g
Synonym Pyrazine-2-carboxylic Acid, Pyrazinamide Imp. A (EP), 2-Pyrazinecarboxylic acid (ACI), Pyrazinecarboxylic acid (7CI','8CI','9CI), Pyrazinoic acid (6CI), 1,4-Diazinecarboxylic acid, 2-Carboxypyrazine, 2-Pyrazinoic acid, NSC 13146, NSC 27192, Pyrazinic acid
InChI Formula InChI=1S/C5H4N2O2/c8-5(9)4-3-6-1-2-7-4/h1-3H,(H,8,9)
SMILES OC(=O)c1cnccn1
IUPAC Name pyrazine-2-carboxylic acid
Molecular Weight (g/mol) 124.1
Formula Weight 124.0273
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.