missing translation for 'onlineSavingsMsg'
Learn More

Pyrazine, TRC

Product Code. 30133585
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133585 10 g 10g
30134098 50 g 50g
30007198 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133585 Supplier Toronto Research Chemicals Supplier No. TRCP84074510G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 290-37-9
Molecular Formula C4 H4 N2
Molecular Weight (g/mol) 80.09
Synonym 1,4-Diazabenzene, 1,4-Diazine, Paradiazine, Piazine, p-Diazine
IUPAC Name pyrazine
SMILES c1cnccn1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C4H4N2/c1-2-6-4-3-5-1/h1-4H
Formula Weight 80.0374
Chemical Name or Material Pyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.