missing translation for 'onlineSavingsMsg'
Learn More

Pyrazine, 99+% (dry wt.) water <1.0%

Product Code. 11491738
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491738 25 g 25g
11481738 5 g 5g
11401748 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491738 Supplier Thermo Scientific Alfa Aesar Supplier No. A13982.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 290-37-9 | C4H4N2 | 80.09 g/mol

Pyrazine is used in flavoring, fragrance, and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrazine is used in flavoring, fragrance, and dyes.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 290-37-9
Molecular Formula C4H4N2
Molecular Weight (g/mol) 80.09
MDL Number MFCD00006122
InChI Key KYQCOXFCLRTKLS-UHFFFAOYSA-N
Synonym 1,4-diazine, p-diazine, paradiazine, piazine, 1,4-diazabenzene, pyrazin, ccris 1331, 1,4-diazin, mixture, mu-pyrazine;
PubChem CID 9261
ChEBI CHEBI:30953
IUPAC Name pyrazine
SMILES C1=CN=CC=N1

Specifications

Melting Point 50°C to 56°C
Density 1.031
Boiling Point 113°C to 116°C
Flash Point 55°C (131°F)
Odor Strong
Refractive Index 1.5235
Quantity 25 g
UN Number UN1325
Beilstein 103905
Sensitivity Hygroscopic
Merck Index 14,7957
Solubility Information Soluble in water.
Formula Weight 80.09
Percent Purity ≥99%
Physical Form Crystals or Powder or Crystalline Powder or Fused Solid
Assay (dry wt.) water <1.0%
Chemical Name or Material Pyrazine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.