missing translation for 'onlineSavingsMsg'
Learn More

Pyrazine-2-thiocarboxamide, 97%

Product Code. 15433537
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15433537 250 mg 250mg
11346809 1 g 1g
11356809 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15433537 Supplier Thermo Scientific Alfa Aesar Supplier No. H26750.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4604-72-2 | C5H5N3S | 139.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in organic solvents, ethanol, DMSO.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4604-72-2
Molecular Formula C5H5N3S
Molecular Weight (g/mol) 139.176
MDL Number MFCD00173663
InChI Key LIURPUMROGYCLW-UHFFFAOYSA-N
Synonym thiopyrazinamide, pyrazinecarbothioamide, pyrazinecarboxamide, thio, pyrazine-2-thiocarboxamide, 2-pyrazinecarbothioamide, pyrazinecarboxylic acid thioamide, 2-pyrazinethiocarboxamide, pyrazine carbothioamide, pyrazine thiocarboxamide, acmc-20ajt5
PubChem CID 2797467
IUPAC Name pyrazine-2-carbothioamide
SMILES C1=CN=C(C=N1)C(=S)N

Specifications

Melting Point 198°C to 200°C
Odor Odorless
Quantity 250 mg
Solubility Information Soluble in organic solvents,ethanol,DMSO.
Formula Weight 139.17
Percent Purity 97%
Chemical Name or Material Pyrazine-2-thiocarboxamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.