missing translation for 'onlineSavingsMsg'
Learn More

Pyrazine-2-carbonitrile, 97+%

Product Code. 11494413
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494413 25 g 25g
11484413 5 g 5g
11404423 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494413 Supplier Thermo Scientific Alfa Aesar Supplier No. A10974.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19847-12-2 | C5H3N3 | 105.10 g/mol

Pyrazine-2-carbonitrile is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrazine-2-carbonitrile is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19847-12-2
Molecular Formula C5H3N3
Molecular Weight (g/mol) 105.10
MDL Number MFCD00049361
InChI Key PMSVVUSIPKHUMT-UHFFFAOYSA-N
Synonym pyrazinecarbonitrile, 2-cyanopyrazine, cyanopyrazine, 2-pyrazinecarbonitrile, pyrazinenitrile, pyrazinonitrile, 2-cyanopyarine, 2-cyano pyrazine
PubChem CID 73172
ChEBI CHEBI:3982
IUPAC Name pyrazine-2-carbonitrile
SMILES N#CC1=CN=CC=N1

Specifications

Melting Point 18°C to 20°C
Density 1.174
Boiling Point 90°C to 93°C (15 mmHg)
Flash Point 96°C (204°F)
Refractive Index 1.536
Quantity 25 g
UN Number UN3276
Beilstein 110008
Solubility Information Slightly soluble in water.
Formula Weight 105.1
Percent Purity ≥97%
Chemical Name or Material Pyrazine-2-carbonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.