missing translation for 'onlineSavingsMsg'
Learn More

Pyrazine-2,5-dicarboxylic acid, 95%

Product Code. p-7056053
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15452159

Brand: Thermo Scientific Alfa Aesar H64795.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 122-05-4 | C6H4N2O4 | 168.11 g/mol

It is an pyrazine used for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an pyrazine used for proteomics research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-05-4
Molecular Formula C6H4N2O4
Molecular Weight (g/mol) 168.11
MDL Number MFCD00216919
InChI Key GMIOYJQLNFNGPR-UHFFFAOYSA-N
Synonym 2,5-pyrazinedicarboxylic acid, 2,5-pyrazinedicarboxylicacid, pubchem8868, 2,5-pyrazine-diacid, ksc177q1r, pyrazinedicarboxylic-2,5 acid
PubChem CID 255280
IUPAC Name pyrazine-2,5-dicarboxylic acid
SMILES OC(=O)C1=CN=C(C=N1)C(O)=O

Specifications

Melting Point 255°C to 260°C (literature)
Quantity 1 g
Beilstein 139985
Solubility Information Slightly soluble in water.
Formula Weight 168.11
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Pyrazine-2,5-dicarboxylic acid
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato