missing translation for 'onlineSavingsMsg'
Learn More

Pyrazinamide, 99%

Product Code. 10561211 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
10g
25g
This item is not returnable. View return policy

Product Code. 10561211

Brand: Thermo Scientific Acros 157641000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98-96-4 | C5H5N3O | 123.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-96-4
Molecular Formula C5H5N3O
Molecular Weight (g/mol) 123.11
MDL Number MFCD00006132
InChI Key IPEHBUMCGVEMRF-UHFFFAOYSA-N
Synonym pyrazinamide, pyrazinecarboxamide, zinamide, pyrazinoic acid amide, 2-pyrazinecarboxamide, aldinamide, aldinamid, pirazinamid, pyrazineamide, pyrafat
PubChem CID 1046
ChEBI CHEBI:45285
IUPAC Name pyrazine-2-carboxamide
SMILES C1=CN=C(C=N1)C(=O)N

Specifications

Melting Point 188°C to 191°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Quantity 100 g
Merck Index 15, 8067
Solubility Information Solubility in water: 15mg/mL. Other solubilities: 13.8mg/mL methanol- 5.7mg/mL abs. ethanol,3.8mg/mL isopropanol- 1.0mg/mL ether,0.01mg/mL isooctane- 7.4mg/mL chloroform
Formula Weight 123.11
Percent Purity 99%
Physical Form Crystalline Powder or Needles
Chemical Name or Material Pyrazinamide, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.