missing translation for 'onlineSavingsMsg'
Learn More

Pyrantel tartrate, MedChemExpress

Product Code. 16455936
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16455936

Brand: Medchem Express HY12641

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pyrantel tartrate, a tetrahydropyrimidine broad-spectrum anthelmintic, and is a nicotinic acetylcholine receptor (nAChR) agonist. Pyrantel tartrate can elicit spastic muscle paralysis in parasitic worms. Pyrantel tartrate can be used for the research of astrointestinal nematodes infections.

Pyrantel tartrate, a tetrahydropyrimidine broad-spectrum anthelmintic, and is a nicotinic acetylcholine receptor (nAChR) agonist. Pyrantel tartrate can elicit spastic muscle paralysis in parasitic worms. Pyrantel tartrate can be used for the research of astrointestinal nematodes infections.

Specifications

Chemical Name or Material Pyrantel tartrate
Physical Form Solid
CAS 33401-94-4
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C15H20N2O6S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 34 mg/mL (95.40 mM)
SMILES CN1CCCN=C1/C=C/C2=CC=CS2.O=C(O)[C@H](O)[C@@H](O)C(O)=O
Molecular Weight (g/mol) 356.39
Formula Weight 356.39
Percent Purity 98.23%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.