missing translation for 'onlineSavingsMsg'
Learn More

Pyrantel Pamoate, TRC

Product Code. 30409141
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30409141

Brand: Toronto Research Chemicals TRCP8405751G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pyrantel pamoate
CAS 22204-24-6
Molecular Formula C23 H16 O6 . C11 H14 N2 S
Quantity 1 g
Synonym Pyrantel embonate, 1-Methyl-2-[(E)-2-(thiophen-2-yl)ethenyl]-1,4,5,6-tetrahydropyrimidine hydrogen 4,4'-methylenebis(3-hydroxynaphthalene-2-carboxylate), 2-Naphthalenecarboxylic acid','4,4'-methylenebis[3-hydroxy-','compd. with 1,4,5,6-tetrahydro-1-methyl-2-[(1E)-2-(2-thienyl)ethenyl]pyrimidine (1:1), 2-Naphthalenecarboxylic acid','4,4'-methylenebis[3-hydroxy-','compd. with (E)-1,4,5,6-tetrahydro-1-methyl-2-[2-(2-thienyl)ethenyl]pyrimidine (1:1), 2-Naphthoic acid','4,4'-methylenebis[3-hydroxy-','compd. with (E)-1,4,5,6-tetrahydro-1-methyl-2-[2-(2-thienyl)vinyl]pyrimidine (1:1) (8CI), Pyrimidine','1,4,5,6-tetrahydro-1-methyl-2-[(1E)-2-(2-thienyl)ethenyl]-','4,4'-methylenebis[3-hydroxy-2-naphthalenecarboxylate] (1:1) (9CI), Pyrimidine','1,4,5,6-tetrahydro-1-methyl-2-[2-(2-thienyl)ethenyl]-','(E)-','4,4'-methylenebis[3-hydroxy-2-naphthalenecarboxylate] (1:1), Antiminth, CP 10423-16, Cobantril, Combantrin, Early Bird, Embovin, Helmex, Helmintox, Nemocid, Piranver, Pyrantel embonate, Pyrantel pamoate
InChI Formula InChI=1S/C23H16O6.C11H14N2S/c24-20-16(14-7-3-1-5-12(14)9-18(20)22(26)27)11-17-15-8-4-2-6-13(15)10-19(21(17)25)23(28)29;1-13-8-3-7-12-11(13)6-5-10-4-2-9-14-10/h1-10,24-25H,11H2,(H,26,27)(H,28,29);2,4-6,9H,3,7-8H2,1H3/b;6-5+
SMILES CN1CCCN=C1\C=C\c2cccs2.OC(=O)c3cc4ccccc4c(Cc5c(O)c(cc6ccccc56)C(=O)O)c3O
IUPAC Name 4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid;1-methyl-2-[(E)-2-thiophen-2-ylethenyl]-5,6-dihydro-4H-pyrimidine
Molecular Weight (g/mol) 594.68
Formula Weight 594.1825
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.