missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrantel Pamoate, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Pyrantel pamoate |
| CAS | 22204-24-6 |
| Molecular Formula | C23 H16 O6 . C11 H14 N2 S |
| Quantity | 250 mg |
| Synonym | Pyrantel embonate, 1-Methyl-2-[(E)-2-(thiophen-2-yl)ethenyl]-1,4,5,6-tetrahydropyrimidine hydrogen 4,4'-methylenebis(3-hydroxynaphthalene-2-carboxylate), 2-Naphthalenecarboxylic acid','4,4'-methylenebis[3-hydroxy-','compd. with 1,4,5,6-tetrahydro-1-methyl-2-[(1E)-2-(2-thienyl)ethenyl]pyrimidine (1:1), 2-Naphthalenecarboxylic acid','4,4'-methylenebis[3-hydroxy-','compd. with (E)-1,4,5,6-tetrahydro-1-methyl-2-[2-(2-thienyl)ethenyl]pyrimidine (1:1), 2-Naphthoic acid','4,4'-methylenebis[3-hydroxy-','compd. with (E)-1,4,5,6-tetrahydro-1-methyl-2-[2-(2-thienyl)vinyl]pyrimidine (1:1) (8CI), Pyrimidine','1,4,5,6-tetrahydro-1-methyl-2-[(1E)-2-(2-thienyl)ethenyl]-','4,4'-methylenebis[3-hydroxy-2-naphthalenecarboxylate] (1:1) (9CI), Pyrimidine','1,4,5,6-tetrahydro-1-methyl-2-[2-(2-thienyl)ethenyl]-','(E)-','4,4'-methylenebis[3-hydroxy-2-naphthalenecarboxylate] (1:1), Antiminth, CP 10423-16, Cobantril, Combantrin, Early Bird, Embovin, Helmex, Helmintox, Nemocid, Piranver, Pyrantel embonate, Pyrantel pamoate |
| InChI Formula | InChI=1S/C23H16O6.C11H14N2S/c24-20-16(14-7-3-1-5-12(14)9-18(20)22(26)27)11-17-15-8-4-2-6-13(15)10-19(21(17)25)23(28)29;1-13-8-3-7-12-11(13)6-5-10-4-2-9-14-10/h1-10,24-25H,11H2,(H,26,27)(H,28,29);2,4-6,9H,3,7-8H2,1H3/b;6-5+ |
| SMILES | CN1CCCN=C1\C=C\c2cccs2.OC(=O)c3cc4ccccc4c(Cc5c(O)c(cc6ccccc56)C(=O)O)c3O |
| IUPAC Name | 4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid;1-methyl-2-[(E)-2-thiophen-2-ylethenyl]-5,6-dihydro-4H-pyrimidine |
| Molecular Weight (g/mol) | 594.68 |
| Formula Weight | 594.1825 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction