missing translation for 'onlineSavingsMsg'
Learn More

Pyrantel pamoate, MedChemExpress

Product Code. 16401646
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16401646

Brand: Medchem Express HY12640

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pyrantel pamoate (Pyrantel embonate), a tetrahydropyrimidine broad-spectrum anthelmintic, is a nicotinic acetylcholine receptor (nAChR) agonist. Pyrantel pamoate can elicit spastic muscle paralysis in parasitic worms. Pyrantel pamoate can be used for the research of astrointestinal nematodes infections.

Pyrantel pamoate (Pyrantel embonate), a tetrahydropyrimidine broad-spectrum anthelmintic, is a nicotinic acetylcholine receptor (nAChR) agonist. Pyrantel pamoate can elicit spastic muscle paralysis in parasitic worms. Pyrantel pamoate can be used for the research of astrointestinal nematodes infections.

Specifications

Chemical Name or Material Pyrantel pamoate
Physical Form Powder
CAS 22204-24-6
Color Yellow
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C34H30N2O6S
For Use With (Application) Neuroscience-Neuromodulation
Synonym Pyrantel embonate
Solubility Information DMSO : 50 mg/mL (84.08 mM; Need ultrasonic)
SMILES O=C(C1=C(O)C(CC2=C3C=CC=CC3=CC(C(O)=O)=C2O)=C4C=CC=CC4=C1)O.CN5CCCN=C5/C=C/C6=CC=CS6
Molecular Weight (g/mol) 594.68
Formula Weight 594.68
Percent Purity 99.94%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.