missing translation for 'onlineSavingsMsg'
Learn More

Pyranine

Product Code. 11387646
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11387646 25 g 25g
11377646 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387646 Supplier Thermo Scientific Alfa Aesar Supplier No. L11252.14
  • ONLINE EXCLUSIVE PRICE
    £123.42
    £100.65 / 25g
    Save £22.77
    18% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6358-69-6 | C16H7Na3O10S3 | 524.37 g/mol

Pyranine is used as a fluorescent pH indicator with emission change in the physiological pH range. It acts as a dye in optical sensor applications. It finds application as a coloring agent and biological stain. It is also used in the measurement of pH of intracellular fluid in the cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyranine is used as a fluorescent pH indicator with emission change in the physiological pH range. It acts as a dye in optical sensor applications. It finds application as a coloring agent and biological stain. It is also used in the measurement of pH of intracellular fluid in the cells.

Solubility
Soluble in water and glacial acetic acid.

Notes
This dye is a highly water soluble, brightly fluorescent dye useful as a fluorescent pH indicator with emission change in the physiological pH range and as a dye in optical sensor applications.Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6358-69-6
Molecular Formula C16H7Na3O10S3
Molecular Weight (g/mol) 524.37
MDL Number MFCD00037575
InChI Key KXXXUIKPSVVSAW-UHFFFAOYSA-K
Synonym pyranine, solvent green 7, c.i. solvent green 7, japan green 204, hpts, FD&C green no. 8, japan green no. 204, pyrene 1, pyranine 120, FD&C green 8
PubChem CID 61388
ChEBI CHEBI:52083
IUPAC Name trisodium;8-hydroxypyrene-1,3,6-trisulfonate
SMILES [Na+].[Na+].[Na+].OC1=CC(=C2C=CC3=C(C=C(C4=CC=C1C2=C34)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Quantity 25 g
Beilstein 4107272
Solubility Information Soluble in water and glacial acetic acid.
Formula Weight 524.39
Chemical Name or Material Pyranine
compliance-icons
Product Identifier
  • Pyranine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Pyranine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.