missing translation for 'onlineSavingsMsg'
Learn More

Pyraclonil, MedChemExpress

Product Code. 16487378
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487378 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16487378 Supplier Medchem Express Supplier No. HY1753110mg
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Pyraclonil is a proporphyrinogen oxidase (PPO) inhibitor. Pyraclonil is a herbicide agent and is highly effective in controlling the susceptible (S) and multiple-herbicide-resistant (MHR) E. indica populations.

Pyraclonil is a proporphyrinogen oxidase (PPO) inhibitor. Pyraclonil is a herbicide agent and is highly effective in controlling the susceptible (S) and multiple-herbicide-resistant (MHR) E. indica populations.

Specifications

Chemical Name or Material Pyraclonil
CAS 158353-15-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H15ClN6
Quantity 10 mg
Solubility Information DMSO : ≥ 30 mg/mL (95.31 mM)
SMILES N#CC1=C(N(C)CC#C)N(C2=NN3CCCCC3=C2Cl)N=C1
Molecular Weight (g/mol) 314.77
Formula Weight 314.77
Percent Purity 98.87%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.