missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyr6, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Pyr6 is a selective inhibitor of TRPC3 with IC50 of 0.49 uM(Ca2+ influx inhibition in thapsigargin depleted native RBL-2H3 cells).
Specifications
Specifications
| Chemical Name or Material | Pyr6 |
| CAS | 245747-08-4 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C17H9F7N4O |
| Quantity | 5 mg |
| Synonym | N-[4-[3,5-Bis(trifluoromethyl)-1H-pyrazol-1-yl]phenyl]-3-fluoro-4-pyridinecarboxamide |
| Solubility Information | DMSO : ≥ 100 mg/mL (239.08 mM) |
| SMILES | O=C(C1=C(F)C=NC=C1)NC2=CC=C(N3N=C(C(F)(F)F)C=C3C(F)(F)F)C=C2 |
| Molecular Weight (g/mol) | 418.27 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction