missing translation for 'onlineSavingsMsg'
Learn More

Pyr3, MedChemExpress

Product Code. p-200095851
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16492986

Brand: Medchem Express HY108465

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pyr3 is a selective inhibitor of transient receptor potential canonical channel 3 (TRPC3), with an IC50 of 700 nM for TRPC3-mediated Ca2+ influx.

Pyr3 is a selective inhibitor of transient receptor potential canonical channel 3 (TRPC3), with an IC50 of 700 nM for TRPC3-mediated Ca2+ influx.

Specifications

Chemical Name or Material Pyr3
Physical Form Solid
CAS 1160514-60-2
Color Off-White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C16H11Cl3F3N3O3
Solubility Information DMSO : ≥ 125 mg/mL (273.74 mM)
SMILES O=C(C1=C(C(F)(F)F)N(C2=CC=C(NC(/C(Cl)=C(Cl)/Cl)=O)C=C2)N=C1)OCC
Molecular Weight (g/mol) 456.63
Formula Weight 456.63
Percent Purity 99.81%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.