missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human IL-17A&IL-17F Heterodimer Protein, His Tag&Strep II Tag

Product Code. 16417769
Click to view available options
Quantity:
1 mg
100 μg
20 μg
Unit Size:
100µg
1mg
20µg

Product Code. 16417769

Brand: ACROBiosystems ILFH52WF1mg

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Interleukin-17A (IL17A) is also known as cytotoxic T-lymphocyte-associated antigen 8 (CTLA8),which is a proinflammatory cytokine produced by activated T cells. IL17A can regulate the activities of NF-kappaB and mitogen-activated protein kinases. Also,IL17A can stimulate the expression of IL6 and cyclooxygenase-2 (PTGS2/COX-2), as well as enhance the production of nitric oxide (NO).Furthermore,IL17A has been found both in glycosylated and nonglycosylated forms. High levels of IL-17 are associated with several chronic inflammatory diseases including rheumatoid arthritis, psoriasis and multiple sclerosis.

Human IL-17A&IL-17F Heterodimer Protein, His Tag&Strep II Tag, produced by co-expression of IL-17A and IL-17F, has a calculated MW of 17.0 kDa (IL-17A) and 18.4 kDa (IL-17F). Subunit IL-17A is fused with a polyhistidine tag at the N-terminus and subunit I

Specifications

Accession Number NP_002181.1 (IL-17 A) and NP_443104.1 (IL-17 F)
Molecular Weight (g/mol) 17.0 kDa (IL-17 A) and 18.4 kDa (IL-17 F)
Quantity 1 mg
Source Human IL-17 A&IL-17; F Heterodimer Protein, His Tag&Strep; II Tag (ILF-H52WF) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Ala 155 (IL-17 A) and Arg 31 - Gln 163 (IL-17 F) (Accession # Q16552-1 (IL-17 A) and Q96PD4-1 (IL-17 F)). Two sequential affinity purification steps were used to ensure exact 1:1 molar ratio of IL17A&IL17F; heterodimer.
Immunogen Gly 24 - Ala 155 (IL-17 A) and Arg 31 - Gln 163 (IL-17 F)
Storage Requirements -20°C
Gene Alias IL-17 A,Interleukin-17 A,CTLA-8,IL-17,IL-17 F,Interleukin-17 F,Cytokine ML-1
Common Name IL-17 A and IL-17 F
Conjugate Unconjugated
Species Human
Protein Tag His Tag and Strep Tag
Buffer PBS, pH7.4
Endotoxin Level Less than 1.0 EU / μg by the LAL method.
Expression System HEK293
Form Powder
Protein Subtype Cytokines and Growth Factors
Purity or Quality Grade >95% as determined by reduced SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.