missing translation for 'onlineSavingsMsg'
Learn More

Pyoluteorin, TRC

Product Code. 18082010
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18082010 2.5 mg 2.5mg
18032140 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18082010 Supplier Toronto Research Chemicals Supplier No. TRCP8403752.5MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25683-07-2
Molecular Formula C11 H7 Cl2 N O3
Molecular Weight (g/mol) 272.0842
Synonym Ketone, 4,5-dichloropyrrol-2-yl 2,6-dihydroxyphenyl (7CI,8CI), Pyoluteorin (6CI), (4,5-Dichloro-1H-pyrrol-2-yl)(2,6-dihydroxyphenyl)methanone, 4,5-Dichloropyrrol-2-yl 2,6-dihydroxyphenylketone, NSC 143092
IUPAC Name (4,5-dichloro-1H-pyrrol-2-yl)-(2,6-dihydroxyphenyl)methanone
SMILES Oc1cccc(O)c1C(=O)c2cc(Cl)c(Cl)[nH]2

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C11H7Cl2NO3/c12-5-4-6(14-11(5)13)10(17)9-7(15)2-1-3-8(9)16/h1-4,14-16H
Formula Weight 270.9803
Percent Purity >95%
Chemical Name or Material Pyoluteorin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.