missing translation for 'onlineSavingsMsg'
Learn More

Pymetrozine, MedChemExpress

Product Code. 16481078
Encompass_Preferred
Click to view available options
Quantity:
250 mg
500 mg
1 g
5 g
Unit Size:
1g
250mg
500mg
5g
This item is not returnable. View return policy

Product Code. 16481078

Brand: Medchem Express HYB0821

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pymetrozine is a feeding inhibitor of Homoptera, in preventing transmission of cauliflower mosaic caulimovirus by the aphid species Myzus persicae (Sulzer).

Pymetrozine is a feeding inhibitor of Homoptera, in preventing transmission of cauliflower mosaic caulimovirus by the aphid species Myzus persicae (Sulzer).

Specifications

Chemical Name or Material Pymetrozine
Physical Form Solid
CAS 123312-89-0
Color Off-White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C10H11N5O
For Use With (Application) COVID-19-anti-virus
Synonym CGA 215944
Solubility Information DMSO : 33.33 mg/mL (153.43 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1NN=C(C)CN1/N=C/C2=CC=CN=C2
Molecular Weight (g/mol) 217.23
Formula Weight 217.23
Percent Purity 99.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.