missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PYK2 Monoclonal Antibody (GT858)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531573
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Mouse (92%), Rat (93%), Bovine (94%). Positive Control: A431, HeLa. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PTK2B (FAK2) is a cytoplasmic tyrosine kinase involved in calcium-induced regulation of ion channels and activation of the map kinase signaling pathway. PTK2B regulates reorganization of the actin cytoskeleton, cell polarization, cell migration, adhesion, spreading, and bone remodeling. FAK2 plays a role in the regulation of the humoral immune response and is required for normal levels of marginal B-cells in the spleen and normal migration of splenic B-cells. It functions in signaling downstream of integrin and collagen receptors, immune receptors, G-protein coupled receptors, cytokine, chemokine, and growth factor receptors, and mediates responses to cellular stress. Aberrant FAK2 expression may play a role in cancer cell proliferation, migration, and invasion in tumor formation and metastasis. Elevated expression has been seen in gliomas, hepatocellular carcinoma, lung cancer, and breast cancer.
Specifications
| PYK2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P70600, Q14289, Q9QVP9 | |
| PTK2B | |
| Recombinant protein encompassing a sequence within the center region of human PYK2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| GT858 | |
| Unconjugated | |
| PTK2B | |
| CADTK; CAK beta; CAKB; CAKbeta; CAK-beta; calcium-dependent tyrosine kinase; Calcium-regulated non-receptor proline-rich tyrosine kinase; cell adhesion kinase beta; cellular adhesion kinase beta; E430023O05Rik; FADK; FADK 2; FADK2; FAK; FAK1; FAK2; focal adhesion kinase 2; FRNK; PKB; pp125FAK; proline-rich tyrosine kinase 2; protein kinase B; protein tyrosine kinase 2 beta; protein-tyrosine kinase 2-beta; PTK; PTK2 protein tyrosine kinase 2 beta; PTK2B; PTK2B protein tyrosine kinase 2 beta; PYK2; RAFTK; related adhesion focal tyrosine kinase | |
| Mouse | |
| Protein A | |
| RUO | |
| 19229, 2185, 50646 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction