missing translation for 'onlineSavingsMsg'
Learn More

PYK2/FAK2 Antibody (GT1132), Novus Biologicals™

Product Code. 18399455 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18399455 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18399455 Supplier Novus Biologicals Supplier No. NBP313573

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PYK2/FAK2 Monoclonal antibody specifically detects PYK2/FAK2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PYK2/FAK2
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GT1132
Conjugate Unconjugated
Dilution Western Blot 0.2-1 ug/ml, Flow Cytometry 1:10-1:1000, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias CADTKCAK-beta, CAK beta, CAKB, Calcium-dependent tyrosine kinase, Cell adhesion kinase beta, EC 2.7.10, FADK2, FAK2EC 2.7.10.2, Focal adhesion kinase 2, PKB, ppyk2, Proline-rich tyrosine kinase 2, protein kinase B, protein tyrosine kinase 2 beta, protein-tyrosine kinase 2-beta, PTK, PTK2B protein tyrosine kinase 2 beta, PYK2Related adhesion focal tyrosine kinase, RAFTKFADK 2
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human PYK2/FAK2. The exact sequence is proprietary.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Cancer, Cellular Markers, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 2185
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.