missing translation for 'onlineSavingsMsg'
Learn More

Pydiflumetofen, TRC

Product Code. 17992145
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17992145 25 mg 25mg
17951205 5 mg 5mg
17944055 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17992145 Supplier Toronto Research Chemicals Supplier No. TRCP99904025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1228284-64-7
Molecular Formula C16 H16 Cl3 F2 N3 O2
Molecular Weight (g/mol) 426.67
Synonym 3-(Difluoromethyl)-N-methoxy-1-methyl-N-[1-methyl-2-(2,4,6-trichlorophenyl)ethyl]-1H-pyrazole-4-carboxamide, 3-(Difluoromethyl)-N-methoxy-1-methyl-N-[1-(2,4,6-trichlorophenyl)propan-2-yl]-1H-pyrazole-4-carboxamide, Adepidyn, Pydiflumetofen
IUPAC Name 3-(difluoromethyl)-N-methoxy-1-methyl-N-[1-(2,4,6-trichlorophenyl)propan-2-yl]pyrazole-4-carboxamide
SMILES CON(C(C)Cc1c(Cl)cc(Cl)cc1Cl)C(=O)c2cn(C)nc2C(F)F

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H16Cl3F2N3O2/c1-8(4-10-12(18)5-9(17)6-13(10)19)24(26-3)16(25)11-7-23(2)22-14(11)15(20)21/h5-8,15H,4H2,1-3H3
Formula Weight 425.0276
Percent Purity >95%
Chemical Name or Material Pydiflumetofen
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.