missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PYDC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570122
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 83%; Dog: 83%; Goat: 83%; Guinea Pig: 85%; Horse: 83%; Human: 100%; Yeast: 77%.
ASC2 associates with PYCARD/ASC and modulates its ability to collaborate with MEFV/pyrin and NLRP3/cryopyrin in NF-kappa-B and pro-caspase-1 activation. It suppresses kinase activity of NF-kappa-B inhibitor kinase (IKK) complex, expression of NF-kappa-B inducible genes and inhibits NF-kappa-B activation by cytokines and LPS.
Specifications
| PYDC1 | |
| Polyclonal | |
| Unconjugated | |
| PYDC1 | |
| ASC2; ASCI; cellular POP1; cPOP1; PAAD-only protein 1; POP1; PYC1; PYD (pyrin domain) containing 1; PYDC1; pyrin domain containing 1; pyrin domain-containing protein 1; Pyrin-only protein 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 260434 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8WXC3 | |
| PYDC1 | |
| synthetic peptide directed towards the N-terminal region of human PYDC1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction