missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PYCR2 Antibody (OTI3D1), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73743APC
This item is not returnable.
View return policy
Description
PYCR2 Monoclonal specifically detects PYCR2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| PYCR2 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| PYCR2 | |
| Full length recombinant protein of human PYCR2 (NP_037460) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Flow Cytometry | |
| OTI3D1 | |
| Flow Cytometry | |
| EC 1.5.1.2, FLJ54750, P5C reductase 2, P5CR 2, P5CR2, pyrroline 5-carboxylate reductase isoform, pyrroline-5-carboxylate reductase 2, pyrroline-5-carboxylate reductase family, member 2 | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 29920.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction