missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PYCR1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57934
This item is not returnable.
View return policy
Description
PYCR1 Polyclonal specifically detects PYCR1 in Human samples. It is validated for Western Blot.
Specifications
| PYCR1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| ARCL2B, EC 1.5.1.2, mitochondrial pyrroline-5-carboxylate reductase 1, P5C, P5C reductase 1, P5CR, P5CR 1, PIG45, PP222, PRO3, proliferation-inducing protein 45, PYCR, pyrroline-5-carboxylate reductase 1, pyrroline-5-carboxylate reductase 1, mitochondrial | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%; Rabbit: 92%; Rat: 92%; Canine: 85%; Mouse: 85%; Zebrafish: 85%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| A6NFM2 | |
| PYCR1 | |
| Synthetic peptides corresponding to PYCR1(pyrroline-5-carboxylate reductase 1) The peptide sequence was selected from the middle region of PYCR1. Peptide sequence RSLLINAVEASCIRTRELQSMADQEQVSPAAIKKTILDKDHLPLELGSPE. | |
| 100 μL | |
| metabolism | |
| 5831 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction