Learn More
Invitrogen™ PYCARD Polyclonal Antibody

Description
A suggested positive control is HL60 cell lysate. PA5-19957 can be used with blocking peptide PEP-0082. The PA5-19957 immunogen is located within the last 50 amino acids of ASC. Predicted molecular ~ 22kD. In Western blot applications, this antibody has been observed to detect a band at: 22 kD. Predicted species reactivity based on immunogen sequence: Mouse (57%), Rat (57%).
Specifications
Specifications
| Antigen | PYCARD |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | PYCARD |
| Gene Accession No. | Q9ULZ3 |
| Gene Alias | 9130417A21Rik; Apoptosis-associated speck-like protein containing a CARD; Asc; CARD5; caspase recruitment domain-containing protein 5; hASC; mASC; MGC10332; Pycard; PYD and CARD domain containing; PYD and CARD domain-containing protein; Target of methylation-induced silencing 1; TMS; TMS1; TMS-1; TNS1 |
| Gene Symbols | PYCARD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.