missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PXR (SXR) Ligand Binding Domain Protein, GST Tag Recombinant Protein

Product Code. 10296403
Click to view available options
Quantity:
50 μg
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10296403

Brand: Invitrogen™ PV4841

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Pregnane X Receptor (Steroid and Xenobiotic Receptor), Ligand Binding Domain (PXR (SXR)-LBD) is a 64.7 kDa recombinant human protein (amino acids 111-434) expressed as a GST fusion protein in baculovirus-infected insect cells. PXR (SXR)-LBD was expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator recruitment or displacement studies. Protein Form: Recombinant, Ligand Binding Domain

This gene product belongs to the nuclear receptor superfamily, members of which are transcription factors characterized by a ligand-binding domain and a DNA-binding domain. The encoded protein is a transcriptional regulator of the cytochrome P450 gene CYP3A4, binding to the response element of the CYP3A4 promoter as a heterodimer with the 9-cis retinoic acid receptor RXR. It is activated by a range of compounds that induce CYP3A4, including dexamethasone and rifampicin. Several alternatively spliced transcripts encoding different isoforms, some of which use non-AUG translation initiation codon, have been described for this gene. Additional transcript variants exist, however, they have not been fully characterized.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75469
Concentration See Label
For Use With (Application) Ligand Binding Assay
Formulation proprietary buffer with no preservative
Gene ID (Entrez) 8856
Name PXR (SXR) Ligand Binding Domain Protein, GST Tag
Quantity 50 μg
Regulatory Status RUO - research use only
Gene Alias BXR; mPXR; Nr1i2; Nuclear receptor subfamily 1 group I member 2; nuclear receptor subfamily 1, group 1, member 2; nuclear receptor subfamily 1, group I, member 2; ONR 1; ONR1; orphan nuclear receptor PAR1; Orphan nuclear receptor PXR; PAR; PAR 1; PAR 2; PAR q; PAR1; PAR2; PARq; pregnane x nuclear receptor variant 2; Pregnane x receptor; pregnane x receptor (nuclear receptor sub family 1, group I, member 2); PRR; PXR; PXR0.1; PXR0.2; PXR1; SAR; Steroid and xenobiotic receptor; SXR
Common Name PXR
Protein Length 111-434
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Baculovirus
Form Liquid
Protein Subtype Hormone Receptors
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.