Learn More
Invitrogen™ PXR Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5116715
Product Code. 17269213
  • £455.00 / 100µL
Description
Positive Samples: Mouse liver, Rat kidney Immunogen sequence: TQPLGVQGLT EEQRMMIREL MDAQMKTFDT TFSHFKNFRL PGVLSSGCEL PESLQAPSRE EAAKWSQVRK DLCSLKVSLQ LRGEDGSVWN YKPPAD.
This gene product belongs to the nuclear receptor superfamily, members of which are transcription factors characterized by a ligand-binding domain and a DNA-binding domain. The encoded protein is a transcriptional regulator of the cytochrome P450 gene CYP3A4, binding to the response element of the CYP3A4 promoter as a heterodimer with the 9-cis retinoic acid receptor RXR. It is activated by a range of compounds that induce CYP3A4, including dexamethasone and rifampicin. Several alternatively spliced transcripts encoding different isoforms, some of which use non-AUG translation initiation codon, have been described for this gene. Additional transcript variants exist, however, they have not been fully characterized.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PXR
Polyclonal
Unconjugated
NR1I2
BXR; mPXR; Nr1i2; Nuclear receptor subfamily 1 group I member 2; nuclear receptor subfamily 1, group 1, member 2; nuclear receptor subfamily 1, group I, member 2; ONR 1; ONR1; orphan nuclear receptor PAR1; Orphan nuclear receptor PXR; PAR; PAR 1; PAR 2; PAR q; PAR1; PAR2; PARq; pregnane X nuclear receptor variant 2; Pregnane X receptor; pregnane X receptor (nuclear receptor sub family 1, group I, member 2); PRR; PXR; PXR.1; PXR.2; PXR1; SAR; Steroid and xenobiotic receptor; SXR
Rabbit
Affinity Chromatography
RUO
18171, 84385, 8856
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
2.87 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
O54915, O75469, Q9R1A7
NR1I2
Recombinant fusion protein containing a sequence corresponding to amino acids 135-230 of human PXR (NP_0038803).
100 μL
Primary
Human, Mouse, Rat