missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PXR Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67912-1-IG-150UL
This item is not returnable.
View return policy
Description
The PXR antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PXR. This antibody recognizes human, mouse, rat antigen. The PXR antibody has been validated for the following applications: WB, IHC, FC (Intra), ELISA analysis.Specifications
| PXR | |
| Monoclonal | |
| 1000 μg/mL | |
| BXR; ONR1; PAR; PAR1; PAR2; PARq; PRR; PXR; SAR; SXR | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 50 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining), ELISA | |
| 1G3A7 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 8856 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction