missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PXMP4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562130
This item is not returnable.
View return policy
Description
Immunogen sequence: FEYHRSTLQP SLQSSMTYLY EDSNVWHDIS DFLVYNKSRP SN Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 88%.
PXMP4 belongs to the peroxisomal membrane protein PXMP2/4 family.
Specifications
| PXMP4 | |
| Polyclonal | |
| Unconjugated | |
| PXMP4 | |
| 24 kDa intrinsic membrane protein; 24 kDa peroxisomal intrinsic membrane protein; 3010018P03Rik; peroxisomal membrane protein 4; peroxisomal membrane protein 4, 24kDa; PMP24; PXMP4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11264 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y6I8 | |
| PXMP4 | |
| Recombinant protein corresponding to Human PXMP4. Recombinant protein control fragment (Product #RP-99322). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction