missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PXMP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118226
This item is not returnable.
View return policy
Description
PXMP4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP
PXMP4 belongs to the peroxisomal membrane protein PXMP2/4 family.
Specifications
| PXMP4 | |
| Polyclonal | |
| Unconjugated | |
| PXMP4 | |
| 24 kDa intrinsic membrane protein; 24 kDa peroxisomal intrinsic membrane protein; 3010018P03Rik; peroxisomal membrane protein 4; peroxisomal membrane protein 4, 24kDa; PMP24; PXMP4 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 11264 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q9Y6I8 | |
| PXMP4 | |
| E. coli-derived Human PXMP4 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction