missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PXMP4 Monoclonal antibody specifically detects PXMP4 in Mouse samples. It is validated for ELISA
Specifications
Specifications
| Antigen | PXMP4 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 7S41 |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | PMP24, PXMP4 peroxisomal membrane protein 4, 24kDa |
| Host Species | Rat |
| Immunogen | Mouse recombinant PXMP4 |
| Purification Method | Protein G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?