missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PXK Polyclonal antibody specifically detects PXK in Human samples. It is validated for Immunoprecipitation
Specifications
Specifications
| Antigen | PXK |
| Applications | Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Immunoprecipitation 8 - 15 ug/mg lysate |
| Formulation | Tris-citrate/phosphate pH 7 -8. |
| Gene Alias | FLJ20335, K-ATPase, K-ATPase long form, Modulator of Na, MONaKA, PX domain containing serine/threonine kinase, PX domain-containing protein kinase-like protein, PX ser/thr kinase v2, PX serine/threonine kinase |
| Host Species | Rabbit |
| Immunogen | Recognizes region between residues Between Between 528 and 578.. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?